Product Details

SNP ID
rs17456
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:155320520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGAGGAAGACAGTTACAGCCAC[A/G]TTATGGCTCTCATTGAACAGTACGC
Phenotype
MIM: 134629
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135821.1 1390 Missense Mutation ATT,GTT I391V NP_001129293.1
NM_001135822.1 1390 Intron NP_001129294.1
NM_001242824.1 1390 Intron NP_001229753.1
NM_001242825.1 1390 Intron NP_001229754.1
NM_002004.3 1390 Missense Mutation ATT,GTT I391V NP_001995.1
XM_005244962.1 1390 Intron XP_005245019.1
XM_005244963.1 1390 Intron XP_005245020.1
Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 1390 Intron NP_001098673.1
NM_001105204.1 1390 Intron NP_001098674.1
NM_001105205.1 1390 Intron NP_001098675.1
NM_001278227.1 1390 Intron NP_001265156.1
NM_001278228.1 1390 Intron NP_001265157.1
NM_001278229.1 1390 Intron NP_001265158.1
NM_001278230.1 1390 Intron NP_001265159.1
NM_014328.4 1390 Intron NP_055143.2
XM_006711254.1 1390 Intron XP_006711317.1
XM_006711256.1 1390 Intron XP_006711319.1
XM_006711257.1 1390 Intron XP_006711320.1
XM_017000891.1 1390 Intron XP_016856380.1
XM_017000892.1 1390 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 1390 UTR 3 NP_001034606.1

View Full Product Details