Product Details

SNP ID
rs8103444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40891756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGAATTTATCGAATCCAGGTCC[A/C]TTATTTTGGAGATGCAGACAGTAAC
Phenotype
MIM: 608054 MIM: 601133
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYP2A7 PubMed Links

Gene Details

Gene
CYP2A7
Gene Name
cytochrome P450 family 2 subfamily A member 7
There are no transcripts associated with this gene.

Gene
CYP2G1P
Gene Name
cytochrome P450 family 2 subfamily G member 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details