Product Details
- SNP ID
-
rs6986660
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:41496002 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGAACCAGAAGTATTTCAGTTTT[G/T]GGGGTTTTTTTAGATTTTGAAATAT
- Phenotype
-
MIM: 609453
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGA7
PubMed Links
Gene Details
- Gene
- GOLGA7
- Gene Name
- golgin A7
View Full Product Details