Product Details

SNP ID
rs4252249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117988494 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAAGTACCTGCTATGAAGTGGC[A/G]CTCCTGAGGTGAGGAAAAGGGAAGA
Phenotype
MIM: 146933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL10RA PubMed Links
Additional Information
For this assay, SNP(s) [rs4252250] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
IL10RA
Gene Name
interleukin 10 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001558.3 257 Silent Mutation GCA,GCG A60A NP_001549.2

View Full Product Details