Product Details
- SNP ID
-
rs4252250
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:117988495 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAAGTACCTGCTATGAAGTGGCG[C/G]TCCTGAGGTGAGGAAAAGGGAAGAG
- Phenotype
-
MIM: 146933
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IL10RA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4252249] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL10RA
- Gene Name
- interleukin 10 receptor subunit alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001558.3 |
258 |
Missense Mutation |
CTC,GTC |
L61V |
NP_001549.2 |
View Full Product Details