Product Details

SNP ID
rs12404355
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:3490537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGGGACTGCCTCTACTAGTGCCT[C/T]GCTGGTCCACCGCTCCGGGATGTGG
Phenotype
MIM: 604266
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF16 PubMed Links

Gene Details

Gene
ARHGEF16
Gene Name
Rho guanine nucleotide exchange factor 16
There are no transcripts associated with this gene.

Gene
MEGF6
Gene Name
multiple EGF like domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001409.3 4479 Silent Mutation GCA,GCG A1539A NP_001400.3
XM_006710406.3 4479 Silent Mutation GCA,GCG A1434A XP_006710469.1
XM_011540885.1 4479 Silent Mutation GCA,GCG A1601A XP_011539187.1
XM_011540886.1 4479 Silent Mutation GCA,GCG A1558A XP_011539188.1
XM_011540887.2 4479 Silent Mutation GCA,GCG A1393A XP_011539189.1
XM_011540888.2 4479 Intron XP_011539190.1
XM_017000533.1 4479 Silent Mutation GCA,GCG A1557A XP_016856022.1

View Full Product Details