Product Details

SNP ID
rs8123062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56494254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGATGTTAAGAACTTTCTGAAGTA[C/T]TTACTTTTAGGGAATGAGGACTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCNT7 PubMed Links

Gene Details

Gene
GCNT7
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080615.1 Intron NP_542182.1
Gene
RTFDC1
Gene Name
replication termination factor 2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283035.1 Intron NP_001269964.1
NM_001283036.1 Intron NP_001269965.1
NM_001283037.1 Intron NP_001269966.1
NM_016407.4 Intron NP_057491.2
XM_017027872.1 Intron XP_016883361.1

View Full Product Details