Product Details

SNP ID
rs1704793
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.15:44961250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACACATCTAATGTAGAGATACA[A/G]ATATAGATATAGATATATTCTAGCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TERB2 PubMed Links

Gene Details

Gene
TERB2
Gene Name
telomere repeat binding bouquet formation protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152448.2 Intron NP_689661.1
XM_011521240.2 Intron XP_011519542.1

View Full Product Details