Product Details

SNP ID
rs4283174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63613344 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGCAAGTTGGGGAGTGCCAAGT[A/G]GGGTGCAACCAGGGGCTGCCTTGAC
Phenotype
MIM: 609088 MIM: 605109
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL22 PubMed Links

Gene Details

Gene
FBXL22
Gene Name
F-box and leucine rich repeat protein 22
There are no transcripts associated with this gene.

Gene
HERC1
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003922.3 Intron NP_003913.3
XM_017022699.1 Intron XP_016878188.1
XM_017022700.1 Intron XP_016878189.1
XM_017022701.1 Intron XP_016878190.1
XM_017022702.1 Intron XP_016878191.1
XM_017022703.1 Intron XP_016878192.1
XM_017022704.1 Intron XP_016878193.1
XM_017022705.1 Intron XP_016878194.1
XM_017022706.1 Intron XP_016878195.1

View Full Product Details