Product Details

SNP ID
rs12902065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66704825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGATTACCTCTGAGAACTGCCCC[C/G]CTTCTTTGTTCTCCTCTTGGGATTT
Phenotype
MIM: 602931
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMAD6 PubMed Links

Gene Details

Gene
SMAD6
Gene Name
SMAD family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005585.4 738 Intron NP_005576.3
XM_011521561.2 738 UTR 5 XP_011519863.1

View Full Product Details