Product Details

SNP ID
rs1709622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184260493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGTGCGAGCCCTCCTGCCAC[A/C]TGAGCCTTCCTCCCTCTGAGTTGAT
Phenotype
MIM: 608721 MIM: 610145
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAMK2N2 PubMed Links

Gene Details

Gene
CAMK2N2
Gene Name
calcium/calmodulin dependent protein kinase II inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033259.2 Intron NP_150284.1
Gene
ECE2
Gene Name
endothelin converting enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037324.2 Intron NP_001032401.1
NM_001100120.1 Intron NP_001093590.1
NM_001100121.1 Intron NP_001093591.1
NM_014693.3 Intron NP_055508.3
NM_032331.3 Intron NP_115707.2

View Full Product Details