Product Details

SNP ID
rs4577407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149477962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAAATTTCACACCTGGTGCTGA[A/G]TAAAAGGCTAAAGACTGGCTAATAT
Phenotype
MIM: 606567
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TM4SF4 PubMed Links

Gene Details

Gene
TM4SF4
Gene Name
transmembrane 4 L six family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004617.3 Intron NP_004608.1

View Full Product Details