Product Details

SNP ID
rs4075389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144112836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACACACCCCTTTCCCCCGTGGAGG[C/T]GCTGGGAGGGCAGGTCCCCTAGGGA
Phenotype
MIM: 610210 MIM: 611885
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAF1 PubMed Links

Gene Details

Gene
MAF1
Gene Name
MAF1 homolog, negative regulator of RNA polymerase III
There are no transcripts associated with this gene.

Gene
SHARPIN
Gene Name
SHANK associated RH domain interactor
There are no transcripts associated with this gene.

Gene
WDR97
Gene Name
WD repeat domain 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316309.1 Intron NP_001303238.1
XM_017013385.1 Intron XP_016868874.1
XM_017013386.1 Intron XP_016868875.1
XM_017013387.1 Intron XP_016868876.1
XM_017013388.1 Intron XP_016868877.1
XM_017013389.1 Intron XP_016868878.1
XM_017013390.1 Intron XP_016868879.1
XM_017013391.1 Intron XP_016868880.1
XM_017013392.1 Intron XP_016868881.1
XM_017013393.1 Intron XP_016868882.1
XM_017013394.1 Intron XP_016868883.1

View Full Product Details