Product Details

SNP ID
rs13135515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1391660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTTTTTTCACCCTTTAACTTTC[A/T]TTGTATCTCTGAATCTAAAGTGTGT
Phenotype
MIM: 610203 MIM: 614632
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CRIPAK PubMed Links

Gene Details

Gene
CRIPAK
Gene Name
cysteine rich PAK1 inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175918.3 109 UTR 5 NP_787114.2
Gene
UVSSA
Gene Name
UV stimulated scaffold protein A
There are no transcripts associated with this gene.

View Full Product Details