Product Details

SNP ID
rs11377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:116154487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATTTCAGCGGGCGAGGCGAGCA[C/G]CTGCGCCTCCGGGCCGACCTCGAGC
Phenotype
MIM: 176385
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PAPPA PubMed Links

Gene Details

Gene
PAPPA
Gene Name
pappalysin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002581.4 736 Missense Mutation CAC,CAG H105Q NP_002572.2
XM_006717129.2 736 Intron XP_006717192.1
XM_017014784.1 736 Missense Mutation CAC,CAG H105Q XP_016870273.1

View Full Product Details