Product Details

SNP ID
rs2661688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78219984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACAGAGGGTTTTTGCCCTTGCAA[A/G]TAGAGCAACTGAAATCTGACACCAT
Phenotype
MIM: 603352
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BIRC5 PubMed Links

Gene Details

Gene
BIRC5
Gene Name
baculoviral IAP repeat containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012270.1 Intron NP_001012270.1
NM_001012271.1 Intron NP_001012271.1
NM_001168.2 Intron NP_001159.2

View Full Product Details