Product Details

SNP ID
rs14451
Assay Type
Functionally tested
NCBI dbSNP Submissions
62
Location
Chr.1:202335740 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
ACATGTGCAGCTCTCTCTTCAGACG[C/T]GAAGCTCTCTGCATGATCCCCAAGT
Phenotype
MIM: 610538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2T PubMed Links

Gene Details

Gene
UBE2T
Gene Name
ubiquitin conjugating enzyme E2 T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310326.1 151 UTR 5 NP_001297255.1
NM_014176.3 151 Silent Mutation NP_054895.1

View Full Product Details