Product Details
- SNP ID
-
rs34239817
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:12573035 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAATGAGAGTACTTGGAAGT[A/G]TTCTTGTGTCAAGAGAGAATTATGG
- Phenotype
-
MIM: 613416
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCIN
PubMed Links
Gene Details
- Gene
- SCIN
- Gene Name
- scinderin
View Full Product Details