Product Details
- SNP ID
-
rs6742619
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:73678464 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATTCCGTAGGGAACACTGGGGCCA[C/T]ATGGTATTACAGTAATATATCATTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALMS1P1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115674385] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALMS1P1
- Gene Name
- ALMS1, centrosome and basal body associated protein pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details