Product Details

SNP ID
rs2712029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:6869051 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCACATGGCAGGATGTGCCTGC[A/G]TATCTAGCCCTTCATATAATGGCAG
Phenotype
MIM: 611787 MIM: 607810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMPK2 PubMed Links

Gene Details

Gene
CMPK2
Gene Name
cytidine/uridine monophosphate kinase 2
There are no transcripts associated with this gene.

Gene
RSAD2
Gene Name
radical S-adenosyl methionine domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080657.4 Intron NP_542388.2
XM_011510414.2 Intron XP_011508716.1
XM_011510415.2 Intron XP_011508717.1

View Full Product Details