Product Details

SNP ID
rs1003968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21013301 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCCTCTAGAAGCACTGCTTCCTA[C/T]GCTCCTTAGGACCACTGCCCGCATA
Phenotype
MIM: 608077
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RX6 PubMed Links

Gene Details

Gene
P2RX6
Gene Name
purinergic receptor P2X 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159554.1 Intron NP_001153026.1
NM_005446.3 Intron NP_005437.2
XM_005261819.3 Intron XP_005261876.1
XM_011530498.2 Intron XP_011528800.1
XM_011530499.2 Intron XP_011528801.1
XM_011530500.2 Intron XP_011528802.1
XM_011530501.2 Intron XP_011528803.1
XM_011530502.2 Intron XP_011528804.1
XM_017029066.1 Intron XP_016884555.1
XM_017029067.1 Intron XP_016884556.1
XM_017029068.1 Intron XP_016884557.1
XM_017029069.1 Intron XP_016884558.1
XM_017029070.1 Intron XP_016884559.1
XM_017029071.1 Intron XP_016884560.1
XM_017029072.1 Intron XP_016884561.1
XM_017029073.1 Intron XP_016884562.1
XM_017029074.1 Intron XP_016884563.1
XM_017029075.1 Intron XP_016884564.1
XM_017029076.1 Intron XP_016884565.1
Gene
THAP7-AS1
Gene Name
THAP7 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TUBA3FP
Gene Name
tubulin alpha 3f pseudogene
There are no transcripts associated with this gene.

View Full Product Details