Product Details

SNP ID
rs4290931
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:181447914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACTATTTTCTCTCTTTATACCT[C/T]TTCTGCACCCTGCCTATAGCCGTAG
Phenotype
MIM: 610083
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TENM3 PubMed Links

Gene Details

Gene
TENM3
Gene Name
teneurin transmembrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080477.3 Intron NP_001073946.1
XM_017008385.1 Intron XP_016863874.1
XM_017008386.1 Intron XP_016863875.1
XM_017008387.1 Intron XP_016863876.1
XM_017008388.1 Intron XP_016863877.1
XM_017008389.1 Intron XP_016863878.1
XM_017008390.1 Intron XP_016863879.1
XM_017008391.1 Intron XP_016863880.1
XM_017008392.1 Intron XP_016863881.1
XM_017008393.1 Intron XP_016863882.1
XM_017008394.1 Intron XP_016863883.1
XM_017008395.1 Intron XP_016863884.1
XM_017008396.1 Intron XP_016863885.1

View Full Product Details