Product Details

SNP ID
rs11804573
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:32657378 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACATGTATATTGTTGACTTCGCC[A/G]TCTCCTGATGTTACTAATTTGAACA
Phenotype
MIM: 602923 MIM: 615891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBBP4 PubMed Links

Gene Details

Gene
RBBP4
Gene Name
RB binding protein 4, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135255.1 Intron NP_001128727.1
NM_001135256.1 Intron NP_001128728.1
NM_005610.2 Intron NP_005601.1
Gene
ZBTB8OS
Gene Name
zinc finger and BTB domain containing 8 opposite strand
There are no transcripts associated with this gene.

View Full Product Details