Product Details

SNP ID
rs6716242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:94793668 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACAATGATTTATTGTCTTCTAG[A/G]TATACCCCTAGTATAGTAATGGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD20A8P PubMed Links

Gene Details

Gene
ANKRD20A8P
Gene Name
ankyrin repeat domain 20 family member A8, pseudogene
There are no transcripts associated with this gene.

View Full Product Details