Product Details

SNP ID
rs4501622
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:1971284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTATTGGCTGGGGGAGGAACAC[A/T]TGAACTCCTAAGGGCTAATACAAGC
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
KBTBD11 PubMed Links

Gene Details

Gene
KBTBD11
Gene Name
kelch repeat and BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014867.2 567 Intron NP_055682.1
XM_011534771.2 567 UTR 5 XP_011533073.1
XM_011534772.2 567 Intron XP_011533074.1
XM_017014114.1 567 Intron XP_016869603.1
XM_017014115.1 567 Intron XP_016869604.1
XM_017014116.1 567 Intron XP_016869605.1
XM_017014117.1 567 Intron XP_016869606.1
Gene
KBTBD11-OT1
Gene Name
KBTBD11 overlapping transcript 1
There are no transcripts associated with this gene.

Gene
LOC101928058
Gene Name
uncharacterized LOC101928058
There are no transcripts associated with this gene.

View Full Product Details