Product Details

SNP ID
rs4595177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5860882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATTACTATGGTGGCTGGTTCCCT[A/T]TAAAAGGGTCATTTCACTGTCCCAC
Phenotype
MIM: 611156
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ERMP1 PubMed Links

Gene Details

Gene
ERMP1
Gene Name
endoplasmic reticulum metallopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024896.2 Intron NP_079172.2
XM_005251587.1 Intron XP_005251644.1
XM_017015139.1 Intron XP_016870628.1
Gene
KIAA2026
Gene Name
KIAA2026
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017969.2 Intron NP_001017969.2
XM_005251385.2 Intron XP_005251442.1
XM_006716730.3 Intron XP_006716793.1
XM_011517759.2 Intron XP_011516061.1
XM_011517760.2 Intron XP_011516062.1
XM_011517761.2 Intron XP_011516063.1
XM_017014331.1 Intron XP_016869820.1
XM_017014332.1 Intron XP_016869821.1
XM_017014333.1 Intron XP_016869822.1
XM_017014334.1 Intron XP_016869823.1
XM_017014335.1 Intron XP_016869824.1
XM_017014336.1 Intron XP_016869825.1

View Full Product Details