Product Details

SNP ID
rs4584218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:112503823 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGATTTTTCTCCCTTCCATTTTCC[C/T]TCCTTCCCTGATCATGACTGTCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1958 PubMed Links

Gene Details

Gene
KIAA1958
Gene Name
KIAA1958
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287036.1 Intron NP_001273965.1
NM_001287038.1 Intron NP_001273967.1
NM_133465.3 Intron NP_597722.1
XM_011518311.2 Intron XP_011516613.1

View Full Product Details