Product Details

SNP ID
rs28607202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:32946018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAAAGATTATAGTTGAAGCATTT[A/T]TATTTTGTGATAGTTTTGCCACTGG
Phenotype
MIM: 609430
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NPAS3 PubMed Links

Gene Details

Gene
NPAS3
Gene Name
neuronal PAS domain protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164749.1 Intron NP_001158221.1
NM_001165893.1 Intron NP_001159365.1
NM_022123.2 Intron NP_071406.1
NM_173159.2 Intron NP_775182.1
XM_005267991.3 Intron XP_005268048.2
XM_005267992.3 Intron XP_005268049.2
XM_011537067.2 Intron XP_011535369.1
XM_011537069.2 Intron XP_011535371.2
XM_011537070.2 Intron XP_011535372.1
XM_011537071.2 Intron XP_011535373.2
XM_011537072.2 Intron XP_011535374.1
XM_017021582.1 Intron XP_016877071.1
XM_017021583.1 Intron XP_016877072.1
XM_017021584.1 Intron XP_016877073.1
XM_017021585.1 Intron XP_016877074.1
XM_017021586.1 Intron XP_016877075.1
XM_017021587.1 Intron XP_016877076.1
XM_017021588.1 Intron XP_016877077.1

View Full Product Details