Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098517.1 | Intron | NP_001091987.1 | |||
NM_001301043.1 | Intron | NP_001287972.1 | |||
NM_001301044.1 | Intron | NP_001287973.1 | |||
NM_001301045.1 | Intron | NP_001287974.1 | |||
NM_014333.3 | Intron | NP_055148.3 | |||
XM_005271494.2 | Intron | XP_005271551.1 | |||
XM_017017457.1 | Intron | XP_016872946.1 | |||
XM_017017458.1 | Intron | XP_016872947.1 | |||
XM_017017459.1 | Intron | XP_016872948.1 | |||
XM_017017460.1 | Intron | XP_016872949.1 | |||
XM_017017461.1 | Intron | XP_016872950.1 |