Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_025004.2 | 816 | Intron | NP_079280.2 | ||
XM_017018346.1 | 816 | Intron | XP_016873835.1 | ||
XM_017018347.1 | 816 | Intron | XP_016873836.1 | ||
XM_017018348.1 | 816 | Intron | XP_016873837.1 | ||
XM_017018349.1 | 816 | UTR 5 | XP_016873838.1 | ||
XM_017018350.1 | 816 | Intron | XP_016873839.1 | ||
XM_017018351.1 | 816 | Intron | XP_016873840.1 | ||
XM_017018352.1 | 816 | Intron | XP_016873841.1 | ||
XM_017018353.1 | 816 | Intron | XP_016873842.1 |