Product Details
- SNP ID
-
rs10833746
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:22338885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCTTTGGTGCCTGCAGAGTTTTT[A/G]TCCTTTACTACACAGGATTTATCTT
- Phenotype
-
MIM: 607563
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC17A6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77408251] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC17A6
- Gene Name
- solute carrier family 17 member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020346.2 |
|
Intron |
|
|
NP_065079.1 |
View Full Product Details