Product Details

SNP ID
rs7939226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74593464 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTGCCTAACCTGTGCAAGTCTT[C/T]TAACTATTGGAAGCTTGACATTTGG
Phenotype
MIM: 611415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLD3 PubMed Links

Gene Details

Gene
POLD3
Gene Name
DNA polymerase delta 3, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006591.2 Intron NP_006582.1
XM_005273716.1 Intron XP_005273773.1
XM_011544734.2 Intron XP_011543036.1

View Full Product Details