Product Details

SNP ID
rs12624609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33703636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGGGCATGAGGAAGGGAGATGG[A/G]GAGGGTGTTTGAGGCTCAGCCCTGC
Phenotype
MIM: 616397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PXMP4 PubMed Links

Gene Details

Gene
PXMP4
Gene Name
peroxisomal membrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007238.4 4633 UTR 3 NP_009169.3
NM_183397.2 4633 UTR 3 NP_899634.1

View Full Product Details