Product Details

SNP ID
rs9671248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77338499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAAAGTTTAACTCTCTCAACCAA[C/T]TGCCAGTCAGGAACTCTTTGATCTA
Phenotype
MIM: 603758
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTZ1 PubMed Links

Gene Details

Gene
GSTZ1
Gene Name
glutathione S-transferase zeta 1
There are no transcripts associated with this gene.

Gene
TMED8
Gene Name
transmembrane p24 trafficking protein family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213601.1 4149 Intron NP_998766.1
XM_017021223.1 4149 UTR 3 XP_016876712.1
XM_017021224.1 4149 UTR 3 XP_016876713.1
XM_017021225.1 4149 UTR 3 XP_016876714.1

View Full Product Details