Product Details

SNP ID
rs34422909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47851836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCTCTGACCTTTCTGGGTGAC[C/T]ACCTCCTCTCCTCCCCTCTCCCAGC
Phenotype
MIM: 608613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SP6 PubMed Links

Gene Details

Gene
SP6
Gene Name
Sp6 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258248.1 Intron NP_001245177.1
NM_199262.2 Intron NP_954871.1
XM_006722115.3 Intron XP_006722178.1

View Full Product Details