Product Details

SNP ID
rs3130284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32172710 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAGCACACAGCATATATGGTGA[C/T]TGTGACAGAACACTCACAGCCATAT
Phenotype
MIM: 600214 MIM: 603099 MIM: 609897 MIM: 603298 MIM: 602677
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGER PubMed Links

Gene Details

Gene
AGER
Gene Name
advanced glycosylation end product-specific receptor
There are no transcripts associated with this gene.

Gene
AGPAT1
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006411.3 Intron NP_006402.1
NM_032741.4 Intron NP_116130.2
XM_005248805.3 Intron XP_005248862.1
XM_005248806.2 Intron XP_005248863.1
XM_011514234.1 Intron XP_011512536.1
Gene
EGFL8
Gene Name
EGF like domain multiple 8
There are no transcripts associated with this gene.

Gene
MIR6721
Gene Name
microRNA 6721
There are no transcripts associated with this gene.

Gene
MIR6833
Gene Name
microRNA 6833
There are no transcripts associated with this gene.

Gene
PPT2
Gene Name
palmitoyl-protein thioesterase 2
There are no transcripts associated with this gene.

Gene
PPT2-EGFL8
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
RNF5
Gene Name
ring finger protein 5
There are no transcripts associated with this gene.

View Full Product Details