Product Details

SNP ID
rs11869224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16621772 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAAAAACACATTTAGGATACTAC[A/G]TATAACCTAGGGATTAAGGAGATCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF624 PubMed Links

Gene Details

Gene
ZNF624
Gene Name
zinc finger protein 624
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020787.3 3206 UTR 3 NP_065838.2
XM_006721562.3 3206 UTR 3 XP_006721625.1
XM_011523970.2 3206 UTR 3 XP_011522272.1

View Full Product Details