Product Details

SNP ID
rs1015077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167113018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAATACTGCGATTCCATTTGTAT[A/G]ACAATCTCGGATGCACTTGTTGTAA
Phenotype
MIM: 601835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCR6 PubMed Links

Gene Details

Gene
CCR6
Gene Name
C-C motif chemokine receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004367.5 Intron NP_004358.2
NM_031409.3 Intron NP_113597.2

View Full Product Details