Product Details

SNP ID
rs34269513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:175177851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGATTTGAAGTATTCTAACTGA[C/T]GTTAGACCTTAAAAACTCAAGACCA
Phenotype
MIM: 123811 MIM: 602736
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATF2 PubMed Links

Gene Details

Gene
ATF2
Gene Name
activating transcription factor 2
There are no transcripts associated with this gene.

Gene
ATP5G3
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002258.4 1700 UTR 3 NP_001002258.1
NM_001190329.1 1700 UTR 3 NP_001177258.1
NM_001689.4 1700 UTR 3 NP_001680.1

View Full Product Details