Product Details

SNP ID
rs12534046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100983770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACCTCATGAGTCGTTATGGAGA[A/T]AACCATAAACGTCCTTTATATGCTT
Phenotype
MIM: 604609
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MUC12 PubMed Links

Gene Details

Gene
MUC12
Gene Name
mucin 12, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164462.1 Intron NP_001157934.1

View Full Product Details