Product Details
- SNP ID
-
rs4368845
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:117677438 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTTTCAATCAGATCTTGAAAGCA[C/T]TGGAATTTCTTTAAAGATGTGAGAA
- Phenotype
-
MIM: 610463
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927919
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78035921] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927919
- Gene Name
- uncharacterized LOC101927919
There are no transcripts associated with this gene.
- Gene
- NUS1
- Gene Name
- NUS1 dehydrodolichyl diphosphate synthase subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138459.3 |
|
Intron |
|
|
NP_612468.1 |
View Full Product Details