Product Details

SNP ID
rs3110696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45915251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGACTTCTCTGCCAAATGTGG[A/G]ACTGAGAACATCCCCGCTGCCTGCG
Phenotype
MIM: 146732
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGFBP3 PubMed Links

Gene Details

Gene
IGFBP3
Gene Name
insulin like growth factor binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000598.4 Intron NP_000589.2
NM_001013398.1 Intron NP_001013416.1
XM_017012152.1 Intron XP_016867641.1

View Full Product Details