Product Details

SNP ID
rs3177555
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:1292391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACGGGAGCCCAGGAGCAGGCAG[T/G]GGGCGCAAGTGCTTTATTCAAAGCG
Phenotype
MIM: 601328
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ACAP3 PubMed Links

Gene Details

Gene
ACAP3
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030649.2 6225 UTR 3 NP_085152.2
XM_005244715.2 6225 UTR 3 XP_005244772.1
XM_005244717.3 6225 UTR 3 XP_005244774.1
XM_011540606.2 6225 UTR 3 XP_011538908.1
XM_011540607.1 6225 UTR 3 XP_011538909.1
XM_011540608.2 6225 UTR 3 XP_011538910.1
XM_011540609.2 6225 UTR 3 XP_011538911.1
XM_017000233.1 6225 UTR 3 XP_016855722.1
Gene
MIR6726
Gene Name
microRNA 6726
There are no transcripts associated with this gene.

Gene
SCNN1D
Gene Name
sodium channel epithelial 1 delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130413.3 6225 Intron NP_001123885.2
XM_011541899.2 6225 Intron XP_011540201.1
XM_011541901.2 6225 Intron XP_011540203.1
XM_011541902.2 6225 Intron XP_011540204.1
XM_011541905.2 6225 Intron XP_011540207.1
XM_011541906.2 6225 Intron XP_011540208.1
XM_011541908.2 6225 Intron XP_011540210.1
XM_011541920.2 6225 Intron XP_011540222.1
XM_011541925.2 6225 Intron XP_011540227.1
XM_011541929.2 6225 Intron XP_011540231.1
XM_011541932.2 6225 Intron XP_011540234.1
XM_011541933.2 6225 Intron XP_011540235.1
XM_017002037.1 6225 Intron XP_016857526.1
XM_017002038.1 6225 Intron XP_016857527.1
XM_017002039.1 6225 Intron XP_016857528.1
XM_017002040.1 6225 Intron XP_016857529.1
XM_017002041.1 6225 Intron XP_016857530.1
XM_017002042.1 6225 Intron XP_016857531.1
XM_017002043.1 6225 Intron XP_016857532.1
XM_017002044.1 6225 Intron XP_016857533.1

View Full Product Details