Product Details

SNP ID
rs3176460
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:50972082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACAATTATGTTTATTTAACTCAA[C/T]TTCAAAAGCCTTTTCCACCTTCCTA
Phenotype
MIM: 603369
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN2C PubMed Links

Gene Details

Gene
CDKN2C
Gene Name
cyclin dependent kinase inhibitor 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001262.2 Intron NP_001253.1
NM_078626.2 Intron NP_523240.1

View Full Product Details