Product Details

SNP ID
rs3180386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73569862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGTGCGCGTGGGCCGGGTGCGA[G/T]GCACGCTCTTCCTGCCGCCAGGTGA
Phenotype
MIM: 609972
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACOT2 PubMed Links

Gene Details

Gene
ACOT2
Gene Name
acyl-CoA thioesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006821.5 778 Intron NP_006812.3
XM_006720007.1 778 Intron XP_006720070.1
XM_017020937.1 778 Missense Mutation GGC,TGC G208C XP_016876426.1
Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 778 Intron NP_001207413.1
NM_203309.2 778 Intron NP_976054.2
XM_006720143.2 778 Intron XP_006720206.1
XM_011536760.2 778 Intron XP_011535062.1
XM_011536761.2 778 Intron XP_011535063.1
XM_017021289.1 778 Intron XP_016876778.1

View Full Product Details