Product Details

SNP ID
rs3204190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91909169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTGCAGCCTGCGAGGAGACCCT[C/T]TGAGGAGCTTTCCTGGGGAAACTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFBP3 PubMed Links

Gene Details

Gene
FGFBP3
Gene Name
fibroblast growth factor binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152429.4 Intron NP_689642.3

View Full Product Details