Product Details

SNP ID
rs3183397
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:180270566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGACTTTAACTAACAGGAACTT[A/G]AGCTTGTTCACTCAAAATCTGTCTA
Phenotype
MIM: 616352 MIM: 602146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACBD6 PubMed Links

Gene Details

Gene
ACBD6
Gene Name
acyl-CoA binding domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032360.3 Intron NP_115736.1
Gene
LHX4
Gene Name
LIM homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033343.3 Intron NP_203129.1
XM_011510105.2 Intron XP_011508407.1
XM_011510106.2 Intron XP_011508408.1
XM_011510108.2 Intron XP_011508410.1
XM_017002755.1 Intron XP_016858244.1
Gene
LHX4-AS1
Gene Name
LHX4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details