Product Details
- SNP ID
-
rs3735430
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:30652370 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTGGGGGCTGGGGCCAGGTATTAG[C/T]CTGGTTGGAGTGATAAGGGACAGAC
- Phenotype
-
MIM: 602034
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRHR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78256172] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRHR2
- Gene Name
- corticotropin releasing hormone receptor 2
View Full Product Details