Product Details

SNP ID
rs3735889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:98194874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGAACACTGAAACAGACTCATG[A/G]TATTAAATAACTGAGCTCCATATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NIPAL2 PubMed Links

Gene Details

Gene
NIPAL2
Gene Name
NIPA like domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321635.1 Intron NP_001308564.1
NM_001321636.1 Intron NP_001308565.1
NM_024759.2 Intron NP_079035.1
XM_011517302.2 Intron XP_011515604.1

View Full Product Details