Product Details

SNP ID
rs3742220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20975125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCCAAGGGCTTTCTTCATCTAC[A/G]GGGTCGAAATTCGAGGTGTCCATGG
Phenotype
MIM: 604861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LATS2 PubMed Links

Gene Details

Gene
LATS2
Gene Name
large tumor suppressor kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014572.2 3492 Silent Mutation CCC,CCT P1004P NP_055387.2
XM_005266342.1 3492 Silent Mutation CCC,CCT P1004P XP_005266399.1
XM_011535042.2 3492 Silent Mutation CCC,CCT P917P XP_011533344.1
XM_017020541.1 3492 Silent Mutation CCC,CCT P897P XP_016876030.1
XM_017020542.1 3492 UTR 3 XP_016876031.1

View Full Product Details